Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009906.5 | NP_034036.1 | 2 - 3 | 110 | 95 | |
GenBank mRNA | AF111172.1 | - | 2 - 3 | 118 | 95 | |
AJ011912.1 | - | 2 - 3 | 104 | 95 | ||
AK002418.1 | - | 2 - 3 | 129 | 95 | ||
AK048279.1 | - | 2 - 3 | 111 | 95 | ||
AK083832.1 | - | 2 - 3 | 116 | 95 | ||
AK146759.1 | - | 2 - 3 | 103 | 95 | ||
AK155021.1 | - | 2 - 3 | 129 | 95 | ||
AK165669.1 | - | 2 - 3 | 135 | 95 | ||
AK169924.1 | - | 2 - 3 | 117 | 95 | ||
AK170781.1 | - | 2 - 3 | 106 | 95 | ||
BC024820.1 | - | 2 - 3 | 112 | 5360558 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |