Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eri2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027698.5 | NP_081974.3 | 2 - 3 | 205 | 93 | |
GenBank mRNA | AK015831.1 | - | 115 | 93 | ||
AK016902.1 | - | 115 | 93 | |||
AK029056.1 | - | 2 - 3 | 130 | 93 | ||
AK135821.1 | - | 2 - 3 | 118 | 93 | ||
AK137621.1 | - | 2 - 3 | 150 | 93 | ||
AK160698.1 | - | 2 - 3 | 101 | 93 | ||
AK165163.1 | - | 2 - 3 | 115 | 93 | ||
BC052150.1 | - | 122 | 93 | |||
BC090961.1 | - | 2 - 3 | 107 | 30648850 | 93 | |
BY019377.1 | - | 2 - 3 | 206 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |