Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016870.3 | NP_058566.3 | 13 - 14 | 1914 | 83 | |
NM_212441.2 | NP_997606.2 | 12 - 13 | 1809 | 83 | ||
NM_212442.2 | NP_997607.2 | 12 - 13 | 1712 | 83 | ||
GenBank mRNA | AB022340.2 | - | 12 - 13 | 1700 | 83 | |
AF068246.1 | - | 12 - 13 | 1699 | 83 | ||
AK077247.1 | - | 12 - 13 | 1775 | 83 | ||
AK143946.1 | - | 1712 | 83 | |||
AK165516.1 | - | 12 - 13 | 1686 | 83 | ||
AY064696.1 | - | 12 - 13 | 1785 | 83 | ||
BC015248.1 | - | 13 - 14 | 1874 | 4239288 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |