Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020575.2 | NP_065600.1 | 4 - 5 | 683 | 109 | |
GenBank mRNA | AF155739.1 | - | 4 - 5 | 683 | 109 | |
AK143177.1 | - | 4 - 5 | 703 | 109 | ||
AK166666.1 | - | 4 - 5 | 690 | 109 | ||
AK167246.1 | - | 4 - 5 | 665 | 109 | ||
AK168555.1 | - | 4 - 5 | 686 | 109 | ||
BC025029.1 | - | 4 - 5 | 647 | 4935124 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |