Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079690.1 | NP_001073158.1 | 24 - 25 | 3183 | 152 | |
NM_183354.2 | NP_899197.2 | 24 - 25 | 3183 | 152 | ||
GenBank mRNA | BC016888.1 | - | 23 - 24 | 3161 | 4236020 | 152 |
BC051100.1 | - | 17 - 18 | 1947 | 152 | ||
U20973.1 | - | 24 - 25 | 3181 | 152 | ||
U20974.1 | - | 24 - 25 | 3181 | 152 | ||
U20975.1 | - | 24 - 25 | 3181 | 152 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |