Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkn1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161624.1 | NP_001155096.1 | 1 - 2 | 197 | 60 | |
GenBank mRNA | U20553.1 | - | 1 - 2 | 30 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus 1700 Probe spans exons |