Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127363.1 | NP_001120835.1 | 5 - 6 | 644 | 109 | |
NM_183144.3 | NP_898967.2 | 5 - 6 | 644 | 109 | ||
GenBank mRNA | AK083449.1 | - | 6 - 7 | 643 | 109 | |
AK157558.1 | - | 5 - 6 | 507 | 109 | ||
AK171360.1 | - | 5 - 6 | 644 | 109 | ||
AK171880.1 | - | 5 - 6 | 620 | 109 | ||
BC056341.1 | - | 5 - 6 | 644 | 5703345 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |