Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025599.3 | NP_001020770.2 | 1 - 2 | 120 | 108 | |
GenBank mRNA | AK142476.1 | - | 1 - 2 | 133 | 108 | |
BC037110.1 | - | 1 - 2 | 102 | 5369645 | 108 | |
CF168373.1 | - | 1 - 2 | 120 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |