Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178404.3 | NP_848491.2 | 3 - 4 | 739 | 134 | |
GenBank mRNA | AK039171.1 | - | 3 - 4 | 734 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |