Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Depdc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144804.1 | NP_659053.1 | 6 - 7 | 1184 | 119 | |
GenBank mRNA | AK134830.1 | - | 6 - 7 | 1204 | 119 | |
AK144066.1 | - | 6 - 7 | 1197 | 119 | ||
AK162086.1 | - | 6 - 7 | 1208 | 119 | ||
AK162218.1 | - | 6 - 7 | 1207 | 119 | ||
AK165504.1 | - | 6 - 7 | 1197 | 119 | ||
BC013499.1 | - | 6 - 7 | 1184 | 4222591 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |