Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rftn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181397.2 | NP_852062.1 | 2 - 3 | 437 | 60 | |
GenBank mRNA | AK082517.1 | - | 50 | 60 | ||
AK087803.1 | - | 210 | 60 | |||
AK152130.1 | - | 2 - 3 | 359 | 60 | ||
AK153288.1 | - | 2 - 3 | 359 | 60 | ||
AK155870.1 | - | 2 - 3 | 401 | 60 | ||
AK170913.1 | - | 2 - 3 | 401 | 60 | ||
AK172884.1 | - | 2 - 3 | 365 | 60 | ||
BC052074.1 | - | 2 - 3 | 437 | 6401192 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |