Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026173.3 | NP_080449.1 | 3 - 4 | 467 | 70 | |
GenBank mRNA | AK004769.2 | - | 3 - 4 | 395 | 70 | |
AK012926.1 | - | 3 - 4 | 435 | 70 | ||
AK089013.1 | - | 3 - 4 | 403 | 70 | ||
AK098149.1 | - | 3 - 4 | 360 | 70 | ||
AK150656.1 | - | 3 - 4 | 468 | 70 | ||
BC016527.1 | - | 3 - 4 | 381 | 3157281 | 70 | |
BC027118.1 | - | 3 - 4 | 405 | 5359153 | 70 | |
BC037631.1 | - | 3 - 4 | 424 | 4954391 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |