Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Baz1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013815.2 | NP_038843.2 | 5 - 6 | 844 | 105 | |
GenBank mRNA | AF033664.1 | - | 4 - 5 | 589 | 105 | |
AK135668.1 | - | 5 - 6 | 844 | 105 | ||
AK166955.1 | - | 2 - 3 | 163 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |