Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019933.2 | NP_064317.2 | 16 - 17 | 1696 | 129 | |
GenBank mRNA | AF106702.1 | - | 15 - 16 | 1524 | 129 | |
AK046683.1 | - | 16 - 17 | 1971 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mus musculus Probe spans exons |