Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045520.3 | NP_001038985.2 | 2 - 3 | 335 | 84 | |
GenBank mRNA | AB093212.1 | - | 1 - 2 | 1422 | 84 | |
AK143544.1 | - | 2 - 3 | 336 | 84 | ||
AK147875.1 | - | 2 - 3 | 320 | 84 | ||
AK168991.1 | - | 2 - 3 | 301 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |