Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045520.3 | NP_001038985.2 | 4 - 5 | 543 | 102 | |
GenBank mRNA | AB093212.1 | - | 3 - 4 | 1630 | 102 | |
AK143544.1 | - | 4 - 5 | 544 | 102 | ||
AK147875.1 | - | 4 - 5 | 528 | 102 | ||
AK168991.1 | - | 4 - 5 | 509 | 102 | ||
BC004080.1 | - | 1 - 2 | 81 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |