Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045520.3 | NP_001038985.2 | 7 - 8 | 1125 | 102 | |
GenBank mRNA | AB093212.1 | - | 6 - 7 | 2212 | 102 | |
AK143544.1 | - | 7 - 8 | 1126 | 102 | ||
AK147875.1 | - | 7 - 8 | 1110 | 102 | ||
AK168991.1 | - | 7 - 8 | 1091 | 102 | ||
BC004080.1 | - | 4 - 5 | 663 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |