Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164056.1 | NP_001157528.1 | 18 - 19 | 2258 | 76 | |
NM_008875.4 | NP_032901.2 | 17 - 18 | 2203 | 76 | ||
GenBank mRNA | AK136111.1 | - | 12 - 13 | 1441 | 76 | |
BC068144.1 | - | 17 - 18 | 2060 | 30532470 | 76 | |
U87868.1 | - | 18 - 19 | 2241 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |