Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164056.1 | NP_001157528.1 | 16 - 17 | 2001 | 104 | |
NM_008875.4 | NP_032901.2 | 15 - 16 | 1946 | 104 | ||
GenBank mRNA | AK136111.1 | - | 10 - 11 | 1184 | 104 | |
BC068144.1 | - | 15 - 16 | 1803 | 30532470 | 104 | |
U87868.1 | - | 16 - 17 | 1984 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |