Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spg20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144988.1 | NP_001138460.1 | 3 - 4 | 1126 | 98 | |
GenBank mRNA | AK035081.1 | - | 2 - 3 | 985 | 98 | |
AK036569.1 | - | 2 - 3 | 449 | 98 | ||
AK129171.1 | - | 3 - 4 | 1105 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |