Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013896.3 | NP_038924.1 | 2 - 3 | 258 | 62 | |
GenBank mRNA | CO809703.1 | - | 189 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |