Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco6c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028942.4 | NP_083218.1 | 11 - 12 | 1972 | 69 | |
GenBank mRNA | AK016647.1 | - | 11 - 12 | 1920 | 69 | |
AK029505.1 | - | 11 - 12 | 1973 | 69 | ||
AK132668.1 | - | 11 - 12 | 1923 | 69 | ||
AK161249.1 | - | 11 - 12 | 1923 | 69 | ||
BC139066.1 | - | 11 - 12 | 1875 | 8862088 | 69 | |
BC139067.1 | - | 11 - 12 | 1875 | 8862089 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |