Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2142575 |
BC099972.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2142575 |
DV659775.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2142575 |
AK132674.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2142575 |
NM_001134741.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2272979 |
AK138828.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1965160 |
DV659775.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1965160 |
NM_001134741.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
120138 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
529241 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
729809 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
920934 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
828213 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
82837 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
890407 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
886769 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
643175 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
478722 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
922839 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
677244 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
574402 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
144255 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
84286 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
212139 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
58665 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
520433 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
225208 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
153561 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
631617 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
601441 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
482954 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
274599 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
121745 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
817101 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
771385 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
785959 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
746601 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
574236 |
AK021368.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
574236 |
XM_001472210.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
574236 |
XM_992447.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
111351 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
46191 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
165902 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
105584 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P169480 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2272979 |
AK138828.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2142575 |
BC099972.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2142575 |
DV659775.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2142575 |
AK132674.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2142575 |
NM_001134741.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1965160 |
DV659775.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1965160 |
NM_001134741.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P169480 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00224540 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04318398 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04318357 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04318293 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P36065 |
BC099972.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P36065 |
DV659775.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P36065 |
AK132674.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P36065 |
NM_001134741.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P390091 |
BC099972.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P390091 |
AY522557.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P390091 |
AK132674.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P390091 |
AK133026.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P390091 |
NM_001134741.1 |
mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P169480 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P04318398 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01141325 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01141317 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01141339 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20742232 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01141315 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20742217 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01141333 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20742172 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20742176 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01141337 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01092663 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01141336 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01092656 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20742173 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20742233 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20742225 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01141320 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01141322 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P20742182 |
- |
- |