Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brms1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037756.2 | NP_001032845.1 | 1 - 2 | 273 | 69 | |
GenBank mRNA | AK154678.1 | - | 1 - 2 | 274 | 69 | |
AK155742.1 | - | 1 - 2 | 274 | 69 | ||
BC034885.1 | - | 1 - 2 | 234 | 1362264 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |