Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gmps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033300.2 | NP_001028472.2 | 4 - 5 | 561 | 145 | |
GenBank mRNA | AK049755.1 | - | 4 - 5 | 525 | 145 | |
AK143197.1 | - | 4 - 5 | 562 | 145 | ||
AK146654.1 | - | 4 - 5 | 579 | 145 | ||
AK167857.1 | - | 4 - 5 | 541 | 145 | ||
AK168239.1 | - | 4 - 5 | 511 | 145 | ||
AK169043.1 | - | 4 - 5 | 545 | 145 | ||
AK169701.1 | - | 4 - 5 | 559 | 145 | ||
BC037232.1 | - | 2 - 3 | 213 | 145 | ||
BC138411.1 | - | 4 - 5 | 517 | 8861431 | 145 | |
BC138412.1 | - | 4 - 5 | 517 | 8861432 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |