Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028664.1 | NP_082940.1 | 4 - 5 | 750 | 105 | |
GenBank mRNA | AK016757.1 | - | 3 - 4 | 669 | 105 | |
BC049713.1 | - | 4 - 5 | 722 | 6741553 | 105 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AK144074.1 | - | Chr. - - [-] on Build | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |