Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025507.2 | NP_079783.2 | 1 - 2 | 50 | 70 | |
GenBank mRNA | AK166163.1 | - | 1 - 2 | 51 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |