Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pip4k2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054051.1 | NP_473392.1 | 1 - 2 | 255 | 80 | |
GenBank mRNA | AK089414.1 | - | 1 - 2 | 245 | 80 | |
AK146559.1 | - | 1 - 2 | 305 | 80 | ||
AK170392.1 | - | 1 - 2 | 182 | 80 | ||
AK171497.1 | - | 1 - 2 | 245 | 80 | ||
AY050219.1 | - | 1 - 2 | 84 | 80 | ||
BC047282.1 | - | 1 - 2 | 255 | 6491155 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |