Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc23l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029430.1 | NP_083706.1 | 5 - 6 | 666 | 65 | |
GenBank mRNA | AK015027.1 | - | 5 - 6 | 638 | 65 | |
BC132384.1 | - | 5 - 6 | 666 | 65 | ||
BC145852.1 | - | 5 - 6 | 666 | 40131155 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |