Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK029745.1 | - | 4 - 5 | 846 | 132 | |
BC116294.1 | - | 4 - 5 | 486 | 40094955 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |