Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033337.4 | NP_001028509.2 | 1 - 2 | 113 | 73 | |
GenBank mRNA | AK078848.1 | - | 1 - 2 | 74 | 73 | |
BC024550.1 | - | 1 - 2 | 63 | 73 | ||
BC030849.1 | - | 1 - 2 | 76 | 73 | ||
BC132621.1 | - | 1 - 2 | 98 | 40130898 | 73 | |
BC132623.1 | - | 1 - 2 | 98 | 40130900 | 73 | |
BY776428.1 | - | 1 - 2 | 114 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |