Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207225.1 | NP_997108.1 | 23 - 24 | 3078 | 81 | |
GenBank mRNA | AK029933.1 | - | 20 - 21 | 2630 | 81 | |
AK155250.1 | - | 24 - 25 | 3264 | 81 | ||
AK162369.1 | - | 1 - 2 | 136 | 81 | ||
BC066052.1 | - | 23 - 24 | 3078 | 6827645 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |