Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027551.2 | NP_081827.1 | 1 - 2 | 60 | 77 | |
GenBank mRNA | AK086025.1 | - | 1 - 2 | 61 | 77 | |
AK146580.1 | - | 1 - 2 | 117 | 77 | ||
AK148398.1 | - | 1 - 2 | 68 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus 1700 Probe spans exons |