Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027551.2 | NP_081827.1 | 4 - 5 | 1135 | 73 | |
GenBank mRNA | AK086025.1 | - | 4 - 5 | 1136 | 73 | |
AK146580.1 | - | 4 - 5 | 1192 | 73 | ||
AK148398.1 | - | 4 - 5 | 1144 | 73 | ||
BC115428.1 | - | 3 - 4 | 1078 | 40094085 | 73 | |
BC115429.1 | - | 3 - 4 | 1078 | 40094088 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |