Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023260.1 | NP_075749.1 | 1 - 2 | 371 | 82 | |
GenBank mRNA | AK002285.1 | - | 1 - 2 | 345 | 82 | |
AK004049.1 | - | 1 - 2 | 335 | 82 | ||
AK007337.1 | - | 1 - 2 | 335 | 82 | ||
AK216319.1 | - | 1 - 2 | 80 | 82 | ||
BC024336.1 | - | 1 - 2 | 330 | 4985502 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |