Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acvrl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277255.1 | NP_001264184.1 | 5 - 6 | 610 | 75 | |
NM_001277257.1 | NP_001264186.1 | 4 - 5 | 549 | 75 | ||
NM_001277258.1 | NP_001264187.1 | 4 - 5 | 518 | 75 | ||
NM_001277259.1 | NP_001264188.1 | 5 - 6 | 605 | 75 | ||
NM_009612.3 | NP_033742.2 | 4 - 5 | 587 | 75 | ||
GenBank mRNA | AK018754.1 | - | 1 - 2 | 89 | 75 | |
AK134712.1 | - | 2 - 3 | 343 | 75 | ||
AK160915.1 | - | 5 - 6 | 578 | 75 | ||
AK170870.1 | - | 2 - 3 | 343 | 75 | ||
BC015083.1 | - | 3 - 4 | 468 | 3980151 | 75 | |
L48015.1 | - | 4 - 5 | 589 | 75 | ||
Z31664.1 | - | 4 - 5 | 555 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons |