Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007742.3 | NP_031768.2 | 31 - 32 | 2193 | 117 | |
GenBank mRNA | AK158011.1 | - | 31 - 32 | 2210 | 117 | |
AK159283.1 | - | 31 - 32 | 2211 | 117 | ||
AK159285.1 | - | 31 - 32 | 2213 | 117 | ||
BC042642.1 | - | 24 - 25 | 1763 | 117 | ||
BC050014.1 | - | 31 - 32 | 2193 | 4225038 | 117 | |
BC059281.1 | - | 31 - 32 | 2203 | 3992363 | 117 | |
M14423.1 | - | 9 - 10 | 543 | 117 | ||
U08020.1 | - | 31 - 32 | 2094 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons |