Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igsf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207205.2 | NP_997088.2 | 6 - 7 | 2342 | 76 | |
GenBank mRNA | AK045721.1 | - | 3 - 4 | 1072 | 76 | |
AK084011.1 | - | 6 - 7 | 2380 | 76 | ||
AK129151.1 | - | 6 - 7 | 2342 | 76 | ||
BC052892.1 | - | 5 - 6 | 1766 | 30055632 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |