Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030096.2 | NP_084372.2 | 7 - 8 | 1025 | 141 | |
GenBank mRNA | AK080767.1 | - | 7 - 8 | 963 | 141 | |
AK133545.1 | - | 7 - 8 | 996 | 141 | ||
AK141200.1 | - | 7 - 8 | 999 | 141 | ||
AK166789.1 | - | 7 - 8 | 951 | 141 | ||
AK195121.1 | - | 3 - 4 | 247 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |