Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027349.3 | NP_081625.3 | 2 - 3 | 304 | 80 | |
GenBank mRNA | AK011403.1 | - | 2 - 3 | 285 | 80 | |
AK151910.1 | - | 2 - 3 | 256 | 80 | ||
AK162135.1 | - | 396 | 80 | |||
AK164367.1 | - | 2 - 3 | 269 | 80 | ||
BC010792.1 | - | 1 - 2 | 50 | 80 | ||
BC158104.1 | - | 1 - 2 | 175 | 9007388 | 80 | |
BY169048.1 | - | 2 - 3 | 306 | 80 | ||
CJ063347.1 | - | 2 - 3 | 297 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |