Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199045.1 | NP_001185974.1 | 2 - 3 | 568 | 123 | |
NM_001199046.1 | NP_001185975.1 | 2 - 3 | 462 | 123 | ||
NM_001199047.1 | NP_001185976.1 | 2 - 3 | 568 | 123 | ||
NM_019756.3 | NP_062730.1 | 2 - 3 | 462 | 123 | ||
GenBank mRNA | AB024329.1 | - | 1 - 2 | 236 | 123 | |
AB031043.1 | - | 1 - 2 | 174 | 123 | ||
AF081568.1 | - | 2 - 3 | 302 | 123 | ||
AK016086.1 | - | 2 - 3 | 262 | 123 | ||
AK030214.1 | - | 2 - 3 | 568 | 123 | ||
AK169454.1 | - | 2 - 3 | 463 | 123 | ||
AK170304.1 | - | 2 - 3 | 427 | 123 | ||
AK171546.1 | - | 2 - 3 | 429 | 123 | ||
BC026465.1 | - | 2 - 3 | 445 | 4484099 | 123 | |
BC132325.1 | - | 2 - 3 | 295 | 40130602 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |