Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134156.2 | NP_598917.1 | 15 - 16 | 2099 | 84 | |
GenBank mRNA | AK142591.1 | - | 6 - 7 | 815 | 84 | |
BC003232.1 | - | 3 - 4 | 344 | 84 | ||
BC023206.1 | - | 2 - 3 | 301 | 84 | ||
BC054830.1 | - | 15 - 16 | 2044 | 6308412 | 84 | |
DQ288940.1 | - | 15 - 16 | 1819 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |