Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164320.1 | NP_001157792.1 | 3 - 4 | 1941 | 71 | |
GenBank mRNA | AK086273.1 | - | 492 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |