Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsbn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172684.2 | NP_766272.2 | 2 - 3 | 1699 | 111 | |
GenBank mRNA | AB101658.1 | - | 2 - 3 | 1701 | 111 | |
AK087685.1 | - | 1 - 2 | 665 | 111 | ||
AK122577.1 | - | 2 - 3 | 1234 | 111 | ||
AK138603.1 | - | 2 - 3 | 1243 | 111 | ||
BC094667.1 | - | 2 - 3 | 1224 | 111 | ||
BC139213.1 | - | 2 - 3 | 1668 | 8862235 | 111 | |
BC139215.1 | - | 2 - 3 | 1668 | 8862237 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |