Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029788.5 | NP_084064.1 | 4 - 5 | 766 | 93 | |
GenBank mRNA | AK002624.1 | - | 4 - 5 | 724 | 93 | |
AK036401.1 | - | 4 - 5 | 767 | 93 | ||
AK153293.1 | - | 4 - 5 | 735 | 93 | ||
AK171077.1 | - | 3 - 4 | 2017 | 93 | ||
BC037057.1 | - | 3 - 4 | 553 | 93 | ||
BC050796.1 | - | 4 - 5 | 734 | 6743413 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |