Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500016792.mm.1 |
NM_001033633.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016792.mm.1 |
NM_001033633.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016792.mm.1 |
TC1500002076.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016789.mm.1 |
NM_001033633.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016789.mm.1 |
NM_001033633.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016789.mm.1 |
TC1500002076.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016785.mm.1 |
NM_001033633.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016785.mm.1 |
NM_001033633.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016785.mm.1 |
TC1500002076.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016788.mm.1 |
NM_001033633.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016788.mm.1 |
NM_001033633.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016788.mm.1 |
TC1500002076.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500016790.mm.1 |
NM_001033633.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016790.mm.1 |
NM_001033633.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500016790.mm.1 |
TC1500002076.mm.1 |
MTA TC |