Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad51c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053269.3 | NP_444499.1 | 1 - 2 | 186 | 77 | |
GenBank mRNA | BC090648.1 | - | 130 | 30640414 | 77 | |
BC141034.1 | - | 1 - 2 | 181 | 9055646 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |