Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008702.2 | NP_001008702.1 | 6 - 7 | 757 | 120 | |
NM_001037905.3 | NP_001032994.1 | 6 - 7 | 831 | 120 | ||
NM_001102400.1 | NP_001095870.1 | 6 - 7 | 831 | 120 | ||
NM_023118.5 | NP_075607.2 | 6 - 7 | 831 | 120 | ||
GenBank mRNA | AK002850.1 | - | 6 - 7 | 714 | 120 | |
AK154716.1 | - | 6 - 7 | 815 | 120 | ||
AK162744.1 | - | 6 - 7 | 802 | 120 | ||
AK162851.1 | - | 6 - 7 | 802 | 120 | ||
AK165519.1 | - | 6 - 7 | 715 | 120 | ||
AK168375.1 | - | 6 - 7 | 805 | 120 | ||
BC006588.1 | - | 5 - 6 | 652 | 3587226 | 120 | |
BC016887.1 | - | 6 - 7 | 697 | 4242174 | 120 | |
CD348957.1 | - | 4 - 5 | 354 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |