Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010408.3 | NP_034538.2 | 2 - 3 | 1195 | 79 | |
GenBank mRNA | AF028737.1 | - | 2 - 3 | 815 | 79 | |
AJ225123.1 | - | 2 - 3 | 1035 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mus musculus Probe spans exons |