Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parvg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162500.1 | NP_001155972.1 | 10 - 11 | 883 | 62 | |
NM_022321.4 | NP_071716.3 | 10 - 11 | 838 | 62 | ||
GenBank mRNA | AF312712.1 | - | 10 - 11 | 796 | 62 | |
AK028329.1 | - | 10 - 11 | 839 | 62 | ||
AK037365.1 | - | 10 - 11 | 811 | 62 | ||
AK040914.1 | - | 10 - 11 | 769 | 62 | ||
AK079881.1 | - | 10 - 11 | 764 | 62 | ||
AK088419.1 | - | 10 - 11 | 788 | 62 | ||
BC011200.1 | - | 10 - 11 | 792 | 4190510 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |